Aminotriester 95%

Supplier: Ambeed
Warning

Synonyms: Di-tert-butyl 4-amino-4-(3-(tert-butoxy)-3-oxopropyl)heptanedioate

A752288-5G A752288-1G
77569-430EA 222.48 USD
77569-430 77573-416
Aminotriester 95%
Aminotriester

Caution: For research use only.

Formula: C₂₂H₄₁NO₆
MW: 415.564 g/mol
MDL Number: MFCD07357283
CAS Number: 136586-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR