Staurosporine 98%

Supplier: Ambeed
Warning

Synonyms: (5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282

A256460-50MG A256460-1G A256460-1MG A256460-250MG A256460-5MG A256460-100MG
77568-484EA 68 USD
77568-484 77668-076 77668-078 77668-080 77668-082 77872-267
Staurosporine 98%
Staurosporine

Caution: For research use only.

Formula: C₂₈H₂₆N₄O₃
MDL Number: MFCD18252446
CAS Number: 62996-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR