2-(Trimethylsilyl)phenol 98%

Supplier: Ambeed
Danger

A1356306-5G
77566-668EA 137 USD
77566-668
2-(Trimethylsilyl)phenol 98%
2-(Trimethylsilyl)phenol

Caution: For research use only.

Formula: C₉H₁₄OSi
MW: 166.2924 g/mol
MDL Number: MFCD09909727
CAS Number: 15288-53-6
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR