Radiant™ Taq DNA Polymerase
Supplier: Alkali Scientific
C101 C105 C109
104028-292EA 166.14 USD
104028-292 104027-250 104026-754
Radiant™ Taq DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Experience reliable amplification with Radiant™ Taq DNA polymerase.
  • Radiant Taq DNA Polymerase, 5u/µl, 500 / 2500/ 10000 units with separate tube 10X PCR buffer and MgCl2
  • High-yields with amplicons up to 5 Kb with standard or fast cycling
  • New-generation PCR buffer formulation including enhancers for maximum PCR efficiency and reaction speed
  • Robust PCR performance across a wide range of DNA templates including genomic DNA and GC-rich and AT-rich sequences
  • Supplied with 10X Reaction Buffer and separate MgCl2 for flexibility and optimization
Radiant™ Taq DNA polymerase description Radiant™ Taq DNA polymerase is a highly purified, high performance DNA polymerase optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA. Radiant™ Taq DNA polymerase exhibits 5´ to 3´ DNA polymerase activity with an error-rate of wild-type Taq (2.0×10-5). The polymerase is ideal for screening, colony PCR, high-throughput PCR and genotyping of problematic GC-rich and AT-rich sequences. Radiant™ Taq DNA polymerase is engineered for robust, superior PCR and is supplied with a highly optimized, new-generation buffer system which provides exceptional sensitivity.
Storage Radiant™ Taq DNA Polymerase is shipped on blue or dry ice and should be stored at –20 °C upon receipt. Excessive freeze/thawing should be avoided. When stored as specified, Radiant™ Taq DNA Polymerase is stable for 12 months from date of receipt. The Kit may also be stored at 4 °C for 1 month. Important Considerations Radiant™ 10X Taq Reaction Buffer: The 10X Taq reaction buffer contains PCR enhancers and has been optimized for maximum efficiency, sensitivity and success with difficult amplicons. We do not suggest the use of additional PCR enhancers. Template: For complex genomic DNA, we suggest the use of 5 to 500 ng per reaction; For cDNA or plasmid DNA, please use <100 ng per reaction.
Primers: Primers should have a predicted melting temperature of around 60 °C, using default Primer 3 settings. The final primer concentration in the reaction should be between 0.2 and 0.6 μM. Annealing: We recommend performing a temperature gradient to determine the optimal annealing temperature. Alternatively, we suggest a 55 °C annealing temperature. Increase in 2 °C increments if non-specific products are present. Extension: Optimal extension is achieved at 72 °C. The optimal extension time is dependent on amplicon length and complexity. 15 seconds per kilobase (Kb) is recommended for amplification from eukaryotic genomic DNA or cDNA.
For shorter amplicons, a 1 second extension is sufficient. Quality control Radiant™ Taq DNA polymerase is tested extensively for robust activity, processivity, efficiency, heat activation, sensitivity, absence of nuclease contamination and absence of nucleic acid contamination. Radiant™ Taq DNA polymerase is manufactured under a comprehensive quality management system, following ISO 9001:2008 standards.
Caution: Limitations of use: This product is intended for research purposes only and is not intended for any animal or human therapeutic use.