Autoclave Sterilization Bags
Supplier: Alkali Scientific
A3000C A3000R A3002C A3002R A3003R A3004R
104026-736EA 116.91 USD
104026-736 104028-788 104029-058 104026-614 104026-656 104028-110
Autoclave Sterilization Bags
Bags Sterilization Pouches
Ensure sterilization excellence with autoclave sterilization bags which ensure that the hazardous waste is properly stored and can be easily transported.
  • Heavy gauge puncture-resistant polypropylene
  • Large white marking area
  • Strong seamless bottom
  • Biohazard symbol clear or red
  • Available in sizes - 12×26", 24×32", 40×46
  • Size: 8.5×11" Packing: 100 per pack
The autoclave sterilization bags and pouches are one of the best products used to isolate and store hazardous waste in various research laboratories. The heavy gauge polyethylene material offers the best protection due to its superior puncture-resistance. This material allows the autoclave sterilization bags and pouches to contain waste at temperatures of up to 136 °C.
Autoclave sterilization bags and pouches have a white marking panel that allows quick and easy to spot labelling. Labelling can be done with any type of marker or permanent marker. The white marking panel is very noticeable due to its color that offers background contrast and also due its size and location.
The words 'Hazardous Waste' and the 'Biohazard' symbol appears on each one of the autoclave sterilization bags and pouches in a visible way. This ensures that accidents are reduced to a minimum and conforms with the international standards for biohazard manipulation - OSHA standard 29CFR1910.1030.
Autoclave sterilization bags and pouches are available in two sizes - the 12×26" and the 24×32", and come in packs of 200 units plus and extra four inner packs of 50 units. The bags and pouches are designed to have high resistance against any type of manipulation due to their seamless bottom, and in the same time to be very flexible.
Manipulating and storing biohazardous waste is an important aspect of any laboratory. To avoid any type of unforeseen and unwanted accidents, high quality sterilization bags are a must. The autoclave sterilization bags and pouches help laboratories by offering:
Resistant and flexible material. The heavy gauge polyethylene is one of the strongest type of polyethylene on the market
Good marking area. The white on red marking area is well placed and large enough to be easily detected by anyone
Seamless bottom. This increases the overall integrity of the autoclave sterilization bags
Visible warnings. The 'Hazardous Waste' words and the 'Biohazard' symbol are clear and large enough to be seen by users