BioBlue-Lite™ Western Blot Films
Supplier: Alkali Scientific
Enhance your Western blotting experiments with the BioBlue-Lite™ Western Blot Film, ensuring reliable and high-quality results for your research.
- Low background, high contrast formula
- Coated on both sides (double emulsion)
- Especially useful with chemiluminescence
- Works well with 14C, 32P, 125I, and 35S
BioBlue-Lite™ Western blot film offers unparalleled performance with its Blu-Lite UHC™ technology, combining high contrast with exceptional sensitivity for autoradiographic and electrophoretic applications. This film is specially designed to deliver a lighter blue background, ensuring optimal visualization of DNA bands with remarkable clarity. Coated on both sides for enhanced consistency and performance, it reliably produces sharp images, making it ideal for various laboratory needs. Packaged in light-safe dispenser boxes with an improved 'lift-off' cover, each Box contains 100 sheets, offering a shelf life of two years from the production date.