BrightStar PVDF Membranes
Supplier: Alkali Scientific
XR730 XR732
104028-634EA 306.14 USD
104028-634 104027-920
BrightStar PVDF Membranes
Transfer Membranes
Employ BrightStar PVDF Membrane measuring 300×3000 mm (30 cm wide × 3.0 meter length) for precise protein transfer applications in your laboratory research.
  • 300×3000 mm (30 cm wide × 3.0m length)
  • Hydrophobic Polyvinylidene Fluoride (PVDF) with 0.22 µm pore size
  • Optimized for binding proteins transferred from a variety of gel matrices
  • Dual tissue paper protected (top and bottom) roll
  • Compared to nitrocellulose, is easier to handle and stain, has increased solvent resistance, and higher signal-to-noise ratio
The BrightStar PVDF Membrane, measuring 300×3000 mm, is meticulously designed for binding proteins transferred from diverse gel matrices. Encased in dual tissue paper protection, both at the top and bottom of the roll, this membrane ensures optimal preservation and handling during storage and transfer processes. Engineered from hydrophobic Polyvinylidene Fluoride (PVDF), it offers superior characteristics compared to nitrocellulose. These include enhanced handling capabilities, improved staining efficiency, heightened solvent resistance, and a superior signal-to-noise ratio. With its e×ceptional performance and reliability, the BrightStar PVDF Membrane is an indispensable tool for protein analysis and research applications in molecular biology laboratories.