BrightStar Nitrocellulose Membranes
Supplier: Alkali Scientific
XR642 XR641
104028-324EA 246.14 USD
104028-324 104027-260
BrightStar Nitrocellulose Membranes
Transfer Membranes
Utilize BrightStar Nitrocellulose Membrane measuring 300×3000 mm, for efficient protein transfer in laboratory experiments.
  • 1 roll
  • 300×3000 mm (30 cm wide × 3.0 m length)
  • 45 µm pore size
  • Compatible with standard immunoblotting and nucleic acid detection methods
  • Superior binding capacity with no background interference
  • Excellent signal strength
The BrightStar™ Nitrocellulose Membrane, sized at 300×3000 mm with a pore size of 0.45 µm, is offered in a single roll format. This membrane is engineered to provide outstanding signal strength coupled with minimal background interference, ensuring precise and reproducible results across a wide range of applications. Its large dimensions make it suitable for various laboratory techniques, including protein and nucleic acid transfer in Western blotting and other molecular biology procedures. With its superior performance and ease of use, this membrane is a valuable asset for researchers and scientists striving for accurate and reliable data.