IVF Culture Dishes
Supplier: Alkali Scientific
VF61 VF60
104026-926EA 513.83 USD
104026-926 104028-452
IVF Culture Dishes
Culture Dishes Petri Dishes
Culture IVF samples effectively with the CellPro™ IVF Culture Dish, providing optimal conditions for cell growth and development.
  • Perfect angle of reservoir well walls for locating and easy washing of samples
  • True flat bottom for maintaining optimal temperature
  • Easy grip design for increased safety
The IVF Culture Dish is a specialized tool designed to support the intricate processes involved in in vitro fertilization (IVF) procedures. Crafted with precision and attention to detail, these culture dishes provide an optimal environment for the cultivation and development of embryos during various stages of the IVF process. Made from high-quality materials such as polystyrene, these dishes offer excellent transparency, allowing for easy visualization and monitoring of embryo growth and morphology under a microscope. The flat, smooth surface ensures uniform cell distribution and adherence, facilitating reliable and consistent results. Each dish is sterile to prevent contamination and maintain the integrity of the cultured embryos. With their specific design tailored to the unique requirements of IVF procedures, IVF Culture Dishes play a crucial role in supporting successful embryo development and subsequent transfer for fertility treatments.