ProCycle™ FlexStrip 0.2 ml PCR 8 Strip Tubes, Individually Attached Flat Caps
Supplier: Alkali Scientific
PC7061
104027-490EA 181.52 USD
104027-490
ProCycle™ FlexStrip 0.2 ml PCR 8 Strip Tubes, Individually Attached Flat Caps
Tubes PCR Tubes
Facilitate sample handling with Alkali Scientific™ ProCycle™ flexStrip 0.2 ml PCR 8-Strip tubes, featuring individually attached flat caps and clear construction, available in packs of 120.
  • 0.2 ml 8-Strip PCR tubes with attached caps (120 strip × 8, 960/pk)
  • Caps are optically clear for qPCR
  • Ultra-thin walls for uniformity and consistency
  • Positive sealing eliminates any evaporation
  • Fits standard spacing on 8-channel pipettes
  • Optical flat caps
  • Certified RNAse, DNAse and pyrogen free
Revolutionize PCR workflows with the ProCycle™ FlexStrip 0.2 ml PCR 8-Strip tubes, meticulously designed to offer unparalleled flexibility and efficiency. Featuring individual-attached flat caps, these tubes ensure rapid thermal transfer and secure sample containment, minimizing the risk of contamination while facilitating effortless handling. With each pack containing 120 tubes, researchers can embark on high-throughput experiments with confidence, knowing that sample integrity and experimental reliability are guaranteed.