CellPro™ Marker Centrifuge Tubes
Supplier: Alkali Scientific
CW5602 CW5603
104028-946EA 138.45 USD
104028-946 104026-712
CellPro™ Marker Centrifuge Tubes
Tubes Centrifuge Tubes
Handle large-volume samples with ease using CellPro™ 50 ml marker centrifuge tubes, designed for robust centrifugation.
  • Sterile
  • RNase, DNase and pyrogen free
  • Polypropylene (PP)
  • White Marker caps
  • Oversized white ID patch on sidewall
CellPro™ 50 ml Marker Centrifuge Tubes feature a unique white Marker screw cap that is easy to write on with a Sharpie or similar lab marker and allows easy identification of sample on the top. They also have easy-to-read graduations and an oversized white ID patch on the sidewall.
The tubes are RNase, DNase and pyrogen free, and come packaged either in Green Lab Certified recyclable sterile racks or resealable sterile bags. CellPro™ Marker Tubes are highly resistant to organic solvents and exhibit a stable sample collection and processing environment for nearly all liquids and solids.