Western Blot Boxes
Supplier: Alkali Scientific
BC1200-19 BC1200-3 BC1200-5 BC1200-7 BC1200-7BK BC1200-9 BC1203 BC1204 BC1206 BC1212 BC1200-11 BC1200-13 BC1200-13BK BC1200-15 BC1200-15BK BC1200-17 BC1300-8BK
104026-920EA 70.75 USD
104026-920 104028-336 104028-518 104028-802 104028-318 104028-660 104028-418 104026-838 104027-024 104027-776 104026-886 104028-774 104027-708 104028-340 104026-662 104027-806 104027-070
Western Blot Boxes
Blotting Apparatus
Protect your Western blots with Alkali Scientific™ Western blot boxes.
  • Perfect for Western blotting
  • Reduce antibody usage
  • Hinged lid minimizes evaporation
  • For halves, wholes, strips, or minis
Western blot Boxes and containers are available in both clear and black for light sensitive samples. They fit nearly any sized blot and are available in common blot sizes and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot Boxes. When you match the size of the Box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.
Priced right and ready for use, these Western blot boxes are the perfect choice for your research.
Ordering information: Not sure which blot Box is right for you? Contact your Avantor representative. Also ask us about other Alkali Scientific® research products and equipment.