Universal 5-way Clamps
Supplier: Heidolph
015892140
10812-592EA 838.77 USD
10812-592
Universal 5-way Clamps
Clamps General Purpose Clamps
The StarFish clamp allows you to hold glassware of virtually any size and comes with a choice of Silicone rubber.
Each clamp features five telescopic arms which can be extended and locked in place to suit your needs. Using different straps on separate clamps allows you to grip the flask neck with the Silicone rubber strap for lifting.