H-Gly-Ala-OH

Supplier: Ambeed

A608001-5G A608001-1G A608001-250MG
77551-824EA 94 USD
77551-824 77669-628 77669-630
H-Gly-Ala-OH
Proteins and Peptides

  • InChI Key: VPZXBVLAVMBEQI-VKHMYHEASA-N


GHS07
Signalword: Warning


Caution: for Research Use Only

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR