Colloidal Oatmeal Coated Latex Gloves
Supplier: NEST SCIENTIFIC
904011 904021 904031
77508-980CS 144.13 USD
77508-980 77508-982 77508-984
Colloidal Oatmeal Coated Latex Gloves
Gloves
Oats extractions is a patented coating recognized by the FDA as a skin protectant, which have been proven by clinical studies could reduce and control rash and dry skin symptoms.
  • Moisturizing: Keep skin moist through its moisture-locking capacity
  • Anti-Oxidation: Protects skin from oxidative damage
  • Protect: Forms an adhesive, closed barrier to protect skin from external stimuli
  • Restoring pH: Maintains skin's protective barrier function by restoring pH balance
  • Soothing: Relieves inflammation, irritation and itchiness and boosts skin's restoration process
  • Clean: Acts as a natural cleanser to remove dead skin cells
Clinical studies have shown that colloidal oatmeal could reduce and control rash, dry skin.
Note: Store in a cool, dry place, and avoid direct sunlight.