PurePoint™ Low Retention Filter Tips, Universal
Supplier: Alkali Scientific
FT1000 FT1010 FT1011 FT1020 FT1050 FT1100 FT1200 FT1250 FT1300 FT95ML
104026-930EA 87.62 USD
104026-930 104027-164 104027-310 104026-694 104027-720 104026-790 104026-964 104027-530 104027-798 104027-224
PurePoint™ Low Retention Filter Tips, Universal
Pipette Tips
Minimize sample retention with Alkali Scientific™ PurePoint™ low retention filter tips, offering universal compatibility for accurate pipetting.
  • Universal-fit for most single and multichannel pipets*
  • Gamma-irradiated sterile and graduated
  • Certified RNase, DNase, Human DNA, pyrogen-free
  • Low-Retention, fine-point design for lowest sample adhesion
  • Ultra-clear polymer withstands organic solvents and provides clear, error-free pipetting
  • Effective hydrophobic aerosol barrier eliminates carry-over and pipettor contamination
Elevate your molecular biology liquid handling tasks with PurePoint™ aerosol barrier filter pipet tips, meticulously crafted using premium-grade 100% polypropylene. Engineered for the most critical applications, these tips offer unparalleled purity and performance. Each tip undergoes gamma irradiation to ensure sterility and is rigorously certified RNase/DNase-Free, human DNA-Free, and pyrogen-free, guaranteeing the integrity of your precious samples.
With their Ultra-Clear construction and Fine-Point design, PurePoint™ aerosol filter tips provide exceptional clarity and precision, facilitating the pipetting of nucleic acid and protein samples with utmost accuracy. Universally compatible with the majority of research-grade pipets, these tips offer seamless integration into your laboratory workflow, allowing you to focus on your research with confidence and peace of mind. Trust PurePoint™ aerosol barrier filter pipet tips for consistent, reliable results in your molecular biology experiments.