PureClean™ Tyvek® 1421B Autoclave Bags with Steam Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
6STK0408 6STK0409 6STK0412 6STK0416 6STK0418 6STK0430 6STK0448 6STK050509 6STK0506 6STK0516 6STK1236 6STK1240 6STK1246 6STK1248 6STK1252 6STK1320 6STK1416 6STK1418 6STK1420 6STK1422 6STK2225 6STK2226 6STK2244 6STK2325 6STK2424 6STK2426 6STK2428 6STK2430 6STK2436 6STK2440 6STK4048 6STK4072 6STK0236 6STK0250 6STK0306 6STK0309 6STK0310 6STK0404 6STK0406 6STK040705 6STK1424 6STK1426 6STK1526 6STK1530 6STK1534 6STK1543 6STK1604 6STK1612 6STK1620 6STK1624 6STK0812 6STK0814 6STK0815 6STK0816 6STK0820 6STK0822 6STK0824 6STK0826 6STK0856 6STK0872 6STK1632 6STK1636 6STK1638 6STK1642 6STK1818 6STK1824 6STK1830 6STK1836 6STK1848 6STK1062 6STK0101 6STK0105 6STK0106 6STK0108 6STK0110 6STK0124 6STK0203 6STK0205 6STK0208 6STK0212 6STK09-512 6STK1010 6STK1014 6STK1015 6STK1020 6STK1022 6STK1024 6STK1030 6STK1034 6STK1040 6STK0522 6STK0526 6STK0606 6STK0608 6STK0610 6STK0612 6STK0638 6STK0662 6STK0710 6STK2442 6STK2448 6STK2460 6STK2462 6STK2472 6STK2628 6STK2638 6STK2818 6STK2834 6STK2838 6STK3246 6STK3250 6STK3412 6STK3434 6STK3636 6STK3642 6STK3646 6STK3648 6STK3660 6STK4040 6STK0806 6STK0808 6STK0809 6STK0810 6STK2840 6STK2844 6STK2864 6STK3016 6STK3020 6STK3030 6STK3036 6STK3040 6STK3048 6STK3060 6STK1212 6STK1216 6STK1218 6STK1220 6STK1222 6STK1224 6STK1227 6STK1232 6STK1234 6STK2020 6STK2022 6STK2024 6STK2026 6STK2028 6STK2030 6STK2040 6STK2044 6STK2222 6STK2224 6STK4872 6STK5050 6STK5054 6STK1855 6STK4242 6STK4272 6STK4416 6STK4650 6STK4836 6STK4848 6STK4860 6STK4864
77530-818CS 2739 USD
77530-818 77530-608 77530-560 77530-668 77530-654 77530-746 77530-806 77530-652 77530-692 77530-764 77530-708 77530-844 77530-858 77530-636 77530-810 77530-658 77530-702 77530-704 77530-866 77530-682 77530-820 77530-622 77530-846 77530-582 77530-780 77530-686 77530-646 77530-862 77530-740 77530-840 77530-598 77530-604 77530-854 77530-782 77530-690 77530-816 77530-742 77530-734 77530-762 77530-834 77530-800 77530-760 77530-868 77530-748 77530-620 77530-562 77530-752 77530-802 77530-774 77530-794 77530-790 77530-546 77530-824 77530-624 77530-542 77530-750 77530-770 77530-576 77530-564 77530-826 77530-640 77530-536 77530-720 77530-588 77530-550 77530-556 77530-674 77530-728 77530-838 77530-628 77530-616 77530-772 77530-758 77530-572 77530-736 77530-768 77530-532 77530-528 77530-836 77530-822 77530-672 77530-732 77530-648 77530-784 77530-698 77530-842 77530-548 77530-852 77530-832 77530-606 77530-632 77530-700 77530-828 77530-726 77530-804 77530-864 77530-738 77530-696 77530-586 77530-642 77530-634 77530-678 77530-808 77530-590 77530-716 77530-786 77530-676 77530-706 77530-712 77530-638 77530-644 77530-554 77530-602 77530-630 77530-660 77530-662 77530-596 77530-814 77530-612 77530-614 77530-870 77530-724 77530-684 77530-580 77530-594 77530-848 77530-688 77530-538 77530-578 77530-618 77530-776 77530-600 77530-872 77530-718 77530-540 77530-666 77530-754 77530-656 77530-756 77530-552 77530-650 77530-584 77530-558 77530-610 77530-722 77530-778 77530-664 77530-710 77530-714 77530-856 77530-860 77530-730 77530-544 77530-694 77530-626 77530-574 77530-566 77530-850 77530-792 77530-592 77530-830 77530-796 77530-670 77530-788
PureClean™ Tyvek® 1421B Autoclave Bags with Steam Indicator
Bags Sterilization Pouches
PureClean™ Autoclave bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® on both sides for maximum breathability during sterilization without the chance of residue deposits on equipment post sterilization
  • Tear and puncture resistant
  • Designed to be heat sealed closed
  • Superior to paper based autoclave pouches and wraps
  • Fast and consistent for equipment preparation area
  • Steam indicator and hydrophobic
  • Lint-free
  • Strong factory seals
  • Exceptional microbial barrier
Standardize and simplify the way equipment is wrapped for steam sterilization. The use of these bags will also save valuable time in the equipment prep area. The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part. Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.Manufactured and packaged in a controlled cleanroom environment.