Cyclopropyl methyl ketone ≥97%

Supplier: Ambeed
Danger

Synonyms: 1-Cyclopropylethanone

A719849-25G A719849-10G
77552-218EA 13 USD
77552-218 77686-716
Cyclopropyl methyl ketone ≥97%
Cyclopropyl methyl ketone

Caution: for Research Use Only

Formula: C₅H₈O
MDL Number: MFCD00001297
CAS Number: 765-43-5
UN: 1224
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR