2,2'-Azanediylbis(propan-1-ol) ≥95%

Supplier: Ambeed
Danger

A574510-1G
77551-724EA 649 USD
77551-724
2,2'-Azanediylbis(propan-1-ol) ≥95%
2,2'-Azanediylbis(propan-1-ol)

Caution: for Research Use Only

Formula: C₆H₁₅NO₂
MW: 133.188 g/mol
MDL Number: MFCD19217215
CAS Number: 2294-46-4
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR