CellPro™ Sodium Chloride, 5 M
Supplier: Alkali Scientific
DB0262 DB0262-CS DS0326 PB0286
104028-332EA 26.14 USD
104028-332 104026-994 104027-668 104027-690
CellPro™ Sodium Chloride, 5 M
Buffers Cell Culture Buffers
Introducing CellPro™ Sodium chloride, meticulously crafted to meet the rigorous demands of molecular biology research.
  • Air tight bottle with tamper-proof seal to ensure quality
  • Sterile
  • Ultra pure
  • Filtered
This molecular biology grade sodium chloride solution ensures exceptional purity and quality, making it ideal for a wide range of laboratory applications, including nucleic acid purification, protein crystallization, and molecular assays. With its precisely formulated concentration and stringent quality control measures, CellPro™ sodium chloride provides researchers with reliable and reproducible results. Trust CellPro™ sodium chloride for your molecular biology experiments, and experience the confidence that comes with superior quality and performance.
Caution: UN No. 3248