Tetrabutylammonium dihydrogen phosphate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate
A240724-5G
A240724-100G
A240724-25G
A240724-500G
A240724-1000G
77550-548EA
6
USD
77550-548
77568-336
77568-338
77600-940
77667-976
Tetrabutylammonium dihydrogen phosphate 98%
Tetrabutylammonium dihydrogen phosphate
Caution: For research use only.
Formula:
C₁₆H₃₈NO₄P MW: 339.46 g/mol |
MDL Number:
MFCD00064526 CAS Number: 5574-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...