1-(Trimethylsilyl)-1-propyne ≥98%

Supplier: Ambeed
Danger

Synonyms: Trimethyl-1-propynylsilane

A229387-25G A229387-100G A229387-10G A229387-5G
77550-484EA 19 USD
77550-484 77593-476 77593-478 77593-480
1-(Trimethylsilyl)-1-propyne ≥98%
1-(Trimethylsilyl)-1-propyne

Caution: for Research Use Only

Formula: C₆H₁₂Si
MW: 112.25
Boiling Pt: 99
Density: 0.759
MDL Number: MFCD00009271
CAS Number: 6224-91-5
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR