Bovine Albumin (BSA), Fraction V
Supplier: RPI
A30075-100.0 A30075-1000.0 A30075-25.0 A30075-250.0 A30075-50.0 A30075-500.0 A30075-5.0
76344-874EA 182.65 USD
76344-874 76344-876 76344-878 76344-880 76344-882 76344-884 77510-176
Bovine Albumin (BSA), Fraction V
Proteins and Peptides
Bovine Albumin (BSA), Fraction V
  • Molecular biology grade
  • IgG free
  • Low bioburden
  • Protease free
  • Solvent free extraction process
  • Manufactured by heat shock process
Caution: Research or further manufacturing use only, not for food or drug use.