SLE Latex Test Kits, ASI, Arlington Scientific
Supplier: ARLINGTON SCIENTIFIC INC.
The ASI SLE Slide Test is a convenient and specific latex slide agglutination assay for the qualitative and semiquantitative detection of anti-deoxyribonucleoprotein (anti-DNP) in human serum. No initial dilution of patient sample is required for this test.
- Proven reliability
- Accurate screening test for SLE
- 1 Minute test
- 2 Liquid controls included
- Proudly made in USA
- Longest shelf life - up to 24 months
- Sensitivity: >86%
- Specificity: >82%
Certifications: CE marked. CPT Code 86225. 510(k)-K821385
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...