ASO Latex Test Kits, ASI
Supplier: ARLINGTON SCIENTIFIC INC.
300025
300050
300100
3001000
80087-268EA
132.82
USD
80087-268
80087-270
80087-264
80087-266
ASO Latex Test Kits, ASI
Clinical Diagnostic Supplies
Clinical Reagents and Kits
The ASI ASO Latex Slide Test is a convenient and specific latex slide agglutination assay for the qualitative and semiquantitative detection of Antistreptolysin-O (ASO) antibodies in human serum. No initial dilution of patient sample is required for this test.
- 2 Minute slide test
- 2 Liquid controls included
- Longest shelf life - up to 18 month
- Proven by AAB, CAP, and API surveys
Certifications: CE Marked. CPT Code 86060. 510(k)-K853857.