N-Vision™ 5600 Series Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
T56555B T56505BL
89032-888EA 24.08 USD
89032-888 89032-898 89032-922 89032-892
N-Vision™ 5600 Series Safety Eyewear, Honeywell Safety
Safety Glasses
These ergonomic safety glasses feature comfortable straight and curved temple ends and allow easy lens replacement.
  • Ergonomic frame is composed of Elasto LT chemical-resistant material
  • With either an universal comfort bridge or a flexi-fit three-point soft nosepiece
Lenses have anti-scratch, anti-UV, and antistatic coatings. Clear, amber, blue, brown, smoke, and vermillion lenses feature antifog coating. I/O mirror lenses are suitable for indoor/outdoor applications.
Certifications: ANSI Z87.1 / CSA Z94.3