Triethanolamine (trolamine) 50% (v/v)

Supplier: Ricca Chemical

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

R8695900-1C
RCR8695900-1CEA 69 USD
RCR8695900-1C
Triethanolamine (trolamine) 50% (v/v)
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 335.4
Melting Pt: 21.6
Flash Pt: 179
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
Merck Index: 13,09739

Order Now

Specification Test Results

Appearance Colorless to pale yellow liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR