RINGERS® R668 Thermal Leather Gloves with Impact and 360° Cut Protection
Supplier: Ansell Healthcare
668-08 668-09 668-10 668-11 668-12 668-13
77528-920CS 1029.83 USD
77528-920 77528-926 77528-900 77528-936 77528-908 77528-912
RINGERS® R668 Thermal Leather Gloves with Impact and 360° Cut Protection
Gloves
Premium leather with 360° cut, level 2 impact, and G-100 Thinsulate™ for cold weather.
  • Thermal goatskin leather, G-100 Thinsulate™ lined for warmth and durability
  • Superior cut-resistant lining for impact gloves with 360° cut resistance
  • Patented TPR design provides knuckles, thumb, and finger-length impact protection
RINGERS® R668 medium-duty impact gloves are fully lined with high-quality goatskin leather and G-100 Thinsulate™ for warmth, granting wearers advanced 360° cut resistance in cold weather environments. Developed with F3 Technology™, these goatskin gloves premium leather formulation and reinforced seams around the index finger and base of the thumb make for highly durable, dexterous personal protection. Their patented high-visibility TPR (thermoplastic rubber) design, covering the fingers and knuckles, ensures excellent impact protection. The slip-on cuff design enables donning and doffing with considerable ease, allowing wearers to find a safe, comfortable fit.
Certifications: ANSI Cut A6; ANSI ABR 4; ANSI/ISEA 105-2006 PUN 3.