Surgical Instrument Cleaner, Integra™ Miltex®
Supplier: Miltex
3-725 3-720
95042-724EA 143.23 USD
95042-724 95042-722
Surgical Instrument Cleaner, Integra™ Miltex®
Cleaning Agents Detergents and Surface Cleansers
A super concentrated formula designed for manual and ultrasonic cleaning, the cleaner can also be used for soaking surgical instruments, accessories, and glassware
The neutral pH formula is biodegradable and rinses cleanly, leaving no residue. It is phoshpate-free, and will not stain instruments. Available in a variety of packaging options.
Ordering information: The 0.24L bottle (95042-722) will produce 32 gallons of solution, and the 3.79L bottle (95042-724) will produce 512 gallons of solution. The dispenser box (95042-726) contains eighty 7.4mL (1/4oz.) packets, and will produce 80 gallons of solution.