Rotors for Centrifuges, 5920 R
Supplier: Eppendorf
5895100007 5895101003 5895117007 5895118003 5895190006
75874-312EA 5412.27 USD
75874-312 75874-314 75875-252 75875-254 75875-256
Rotors for Centrifuges, 5920 R
Centrifuges Rotors
Eppendorf Rotors for centrifuges 5920 R - precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Experience precision in refrigerated large-capacity centrifugation with Eppendorf rotors designed for centrifuges 5920 R. These rotors offer a range of capacities and speeds, providing flexibility for various sample processing needs. Accessorize with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-throughput applications, the centrifuges 5920 R is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your refrigerated large-capacity centrifugation experience.