4-tert-Butylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-tert-Butylbenzeneboronic acid(4-(tert-Butyl)phenyl)boronic acid

12321-25G
77498-038EA 62.5 USD
77498-038
4-tert-Butylphenylboronic acid ≥97%
4-tert-Butylphenylboronic acid

Caution: for Research Use Only

Formula: C₁₀H₁₅BO₂
MW: 178.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01009697
CAS Number: 123324-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR