Silver, annular sputtering target, Diameter 117×89 mm (OD×ID)

Supplier: Electron Microscopy Sciences

91030-AG
102099-784EA 1027.15 USD
102099-784
Silver, annular sputtering target, Diameter 117×89 mm (OD×ID)
Silver

Metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.

Formula: Ag
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR