LifeASSURE™ BFS Filter Cartridges
Supplier: 3M Healthcare
BFS020A01BA BFS020A01FA BFS020A02BA BFS020A02FA BFS020A03BA BFS020A03FA BFS020A01BA1 BFS020A01BK1 BFS020A01CA1 BFS020A01CK1 BFS020A01FA1 BFS020A01FB1 BFS020A01FC1 BFS020A01FK1 BFS020A02BA1 BFS020A02BK1 BFS020A02CA1 BFS020A02CC1 BFS020A02CK1 BFS020A02FA1 BFS020A02FC1 BFS020A03BA1 BFS020A03CA1 BFS020A03FA1
77448-052EA 2229.33 USD
77448-052 77448-046 77448-054 77448-048 77448-056 77448-050 77552-900 77552-902 77552-904 77552-906 77552-908 77552-910 77552-912 77552-914 77552-916 77552-918 77552-920 77552-922 77552-924 77552-926 77552-928 77552-930 77552-932 77552-934
LifeASSURE™ BFS Filter Cartridges
Filters Filter Cartridges
3M™ LifeASSURE™ BFS series filter cartridges are used in critical process stages where sterile/particulates filtration are required. The hydrophobic PTFE membrane in an all polypropylene construction provides excellent chemical resistance and high efficiency contaminant reduction. After manufacture and assembly, cartridges are rinsed with 18 meg ohm-cm DI water in a clean environment under an ISO-certified quality system to provide downstream cleanliness and then double-bagged. No pre-wetting is required for solvent based chemicals.
  • Excellent air flow for fermenter aeration and smaller, economical vent filter assemblies
  • Hydrophobic PTFE membrane that will not become blocked with moisture
  • Provides a low differential pressure to minimize bubble formation pressure applications
  • Increased throughput and long filter life
For food and beverage applications (with reinforced stainless steel ring):
Sterile vent filtration for fermentation applications
Sterile air in-take for bioreactors
Sterile compressed gas filtration
Spirit particulate filtration
For microelectronics applications (with embedded polyethersulfone (PES) ring):
High purity chemicals
Organic solvents
Photoresists
Anti-reflective coatings
Plating solutions
Certifications: ISO 9001 Registered quality system, USA FDA CFR-21 compliant materials of construction.