2,3,5,6-Tetrafluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,3,5,6-Tetrafluorophenyl)metaboric acid, (2,3,5,6-Tetrafluorophenyl)boronic acid, Dihydroxy(2,3,5,6-tetrafluorophenyl)borane, 2,3,5,6-Tetrafluorobenzeneboronic acid

12694-5G 12694-25G
77487-344EA 161.71 USD
77487-344 77498-148
2,3,5,6-Tetrafluorophenylboronic acid ≥97%
2,3,5,6-Tetrafluorophenylboronic acid

Caution: For research use only

Formula: C₆H₃BF₄O₂
MW: 193.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039310
CAS Number: 511295-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR