Low Density Flat Poly Bags, Associated Bag
Supplier: Associated Bag
28-4-639 28-5-536 28-5-547 28-5-557 28-5-554 28-6-799 28-6-697 28-4-514 28-6-669 28-6-635 28-6-695 28-6-646 28-6-850 28-6-606 28-6-610 28-6-624 28-6-691 28-6-769 28-6-968 28-4-566 28-4-698
11215-930CS 162.22 USD
11215-930 47735-416 47735-418 47735-420 47735-422 47743-777 47745-020 47746-468 80086-346 47743-773 47743-775 80086-344 80087-528 80086-338 80086-340 80086-342 80086-348 80087-526 47743-779 10030-710 10030-712
Low Density Flat Poly Bags, Associated Bag
Bags Storage Bags
Economical and reusable.
2 to 8 mil for light- to super-heavy weight uses. Not recommended for liquids.
Certifications: Meets FDA and USDA specifications for food contact.