2-Fluoro-5-nitro-4-(trifluoromethyl)benzaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
98796-1G
98796-500MG
98796-5G
77473-944EA
766.32
USD
77473-944
77486-556
77497-178
2-Fluoro-5-nitro-4-(trifluoromethyl)benzaldehyde ≥95%
2-Fluoro-5-nitro-4-(trifluoromethyl)benzaldehyde
Caution: For research use only
Formula:
C₈H₃F₄NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD34819597 CAS Number: 2383649-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...