2-Chloro-5-(dimethylcarbamoyl)phenylboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
55075-1G
55075-500MG
55075-5G
77471-884EA
534.57
USD
77471-884
77484-520
77495-040
2-Chloro-5-(dimethylcarbamoyl)phenylboronic acid ≥95%
2-Chloro-5-(dimethylcarbamoyl)phenylboronic acid
Caution: For research use only
Formula:
C₉H₁₁BClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22125080 CAS Number: 1449144-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...