4-Fluoro-5-methyl-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53560-500MG 53560-5G 53560-25G
77484-444EA 296.64 USD
77484-444 77494-954 77504-434
4-Fluoro-5-methyl-2-(methylthio)benzaldehyde ≥97%
4-Fluoro-5-methyl-2-(methylthio)benzaldehyde

Caution: For research use only

Formula: C₉H₉FOS
Storage Temperature: Ambient
MDL Number: MFCD34757074
CAS Number: 2092702-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR