2-Bromo-4-methylthiophene-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

45931-500MG 45931-5G
77483-972EA 519.12 USD
77483-972 77494-440
2-Bromo-4-methylthiophene-3-carboxylic acid ≥97%
2-Bromo-4-methylthiophene-3-carboxylic acid

Caution: For research use only

Formula: C₆H₅BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD22036015
CAS Number: 1266396-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR