1-(6-Chloro-2-fluoro-3-methoxyphenyl)butan-1-ol ≥95%
Supplier: AOBChem USA
372061-5G
372061-25G
77493-820EA
1484.23
USD
77493-820
77503-562
1-(6-Chloro-2-fluoro-3-methoxyphenyl)butan-1-ol ≥95%
1-(6-Chloro-2-fluoro-3-methoxyphenyl)butan-1-ol
Caution: For research use only
Formula:
C₁₁H₁₄ClFO₂ |
CAS Number:
3002515-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...