1-(4-butylphenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

367732-5G 367732-25G
77493-746EA 924.94 USD
77493-746 77503-500
1-(4-butylphenyl)-2-methylpropan-1-one ≥95%
1-(4-butylphenyl)-2-methylpropan-1-one

Caution: For research use only

Formula: C₁₄H₂₀O
CAS Number: 873962-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR