(6-Chloro-2-fluoro-3-(methylthio)phenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

367691-5G 367691-25G
77493-742EA 1484.23 USD
77493-742 77503-496
(6-Chloro-2-fluoro-3-(methylthio)phenyl)(ethyl)sulfane ≥95%
(6-Chloro-2-fluoro-3-(methylthio)phenyl)(ethyl)sulfane

Caution: For research use only

Formula: C₉H₁₀ClFS₂
CAS Number: 3002488-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR