(3-Bromo-2-chloro-6-fluorophenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

365232-5G 365232-25G
77493-602EA 1484.23 USD
77493-602 77503-366
(3-Bromo-2-chloro-6-fluorophenyl)(cyclopropyl)methanol ≥95%
(3-Bromo-2-chloro-6-fluorophenyl)(cyclopropyl)methanol

Caution: For research use only

Formula: C₁₀H₉BrClFO
CAS Number: 3002532-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR