1-(4-butoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

346823-1G 346823-500MG 346823-5G 346823-25G
77470-040EA 253.38 USD
77470-040 77482-708 77493-136 77502-944
1-(4-butoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-butoxyphenyl)-2,2,2-trifluoroethanone

Caution: For research use only

Formula: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
CAS Number: 159835-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR