1-(2,4-dimethoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

320469-1G 320469-500MG 320469-5G
77469-172EA 391.4 USD
77469-172 77481-848 77492-248
1-(2,4-dimethoxyphenyl)propan-2-ol ≥95%
1-(2,4-dimethoxyphenyl)propan-2-ol

Caution: For research use only

Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
CAS Number: 1014-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR