3-(3-Bromo-2,6-difluorophenyl)tetrahydro-3-furanol ≥97%

Supplier: AOBChem USA

Warning

268545-1G 268545-500MG 268545-5G 268545-25G
77467-856EA 340.93 USD
77467-856 77480-556 77490-896 77500-956
3-(3-Bromo-2,6-difluorophenyl)tetrahydro-3-furanol ≥97%
3-(3-Bromo-2,6-difluorophenyl)tetrahydro-3-furanol

Caution: For research use only

Formula: C₁₀H₉BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34878106
CAS Number: 1702069-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR