2-(4-Brom-3-chloro-2-fluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

32039-1G 32039-500MG 32039-5G
77469-166EA 697.31 USD
77469-166 77481-842 77492-242
2-(4-Brom-3-chloro-2-fluorophenyl)-1,3-dioxolane ≥95%
2-(4-Brom-3-chloro-2-fluorophenyl)-1,3-dioxolane

Caution: For research use only

Formula: C₉H₇BrClFO₂
MDL Number: MFCD31619436
CAS Number: 2221812-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR