(3-(Benzyloxy)phenyl)triethylsilane ≥95%

Supplier: AOBChem USA

319820-1G 319820-500MG 319820-5G 319820-25G
77469-110EA 415.09 USD
77469-110 77481-786 77492-184 77502-116
(3-(Benzyloxy)phenyl)triethylsilane ≥95%
(3-(Benzyloxy)phenyl)triethylsilane

Caution: For research use only

Formula: C₁₉H₂₆OSi
CAS Number: 3002524-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR