Two-Tone® Neoprene Natural Rubber Gloves, Mapa Professionnel
Supplier: MAPA
405427ZQK 405428ZQK 405429ZQK 405420ZQK
32917-200CS 274.79 USD
32917-200 32917-203 32917-206 32917-209 32917-212
Two-Tone® Neoprene Natural Rubber Gloves, Mapa Professionnel
Gloves
Two-tone NS-53 flock-lined, heavyweight gloves offer economical, outstanding protection for chemical processing, food processing, cleaning parts with MEK, aircraft maintenance, fiberglass layup, and general maintenance.
  • Recessed diamond-grip pattern provides a nonslip grip, even on hard-to-grasp materials
  • Two-color construction ensures visual indicator of glove wear-through
  • Yellow, soft, natural rubber provides flexibility and comfort
  • Cotton flock lining absorbs perspiration and enables easy on/off
  • Flat cuff
  • Hand-specific
Double-dipped, blue neoprene-blend safety zone reinforces work area strength and chemical resistance. Gloves resist acids, alcohol, oils, grease, alkalis, caustics, salts, ketones, petroleum derivatives, and solvents.
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.
Certifications: In compliance with CFR 21 170-199.
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.