Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 450)

Supplier: AAT BIOQUEST INC
10033040 10033041
77021-736EA 244.24 USD
77021-736 77022-954
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 450)
Antibodies
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR